Ampac™ Flexibles SealPAK Pouches
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Supplier:  Ampac™ 40224
 View more versions of this product
Catalog No. 01-812-16
Explore available promotions
Description
Description
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Recommended for applications requiring long-term storage, sharper objects, or chemical solutions.
  • Transparent pouches with polyethylene lining
  • Can withstand infrared and gamma rays
  • May be frozen, microwaved, or boiled without harm
  • Easily resealable
  • 2.5 mil
  • Use with Ampac Flexibles pouch sealer (Cat. Nos. 01-812-15 or 01-812-15B) and handy pouch cutter (01-812-21)
Order Info
Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.
Specifications
Specifications
16 oz.
Polyester
-56°C to +115°C
8 x 6.5 in. (20.3 x 16.5cm)
20.3 cm
16.5 cm
Transparent
1800/Cs.
2.50 mil
8 in.
6.5 in.
Videos
SDS
Documents
Documents
Accessories