Ampac™ Flexibles SealPAK Pouches
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Supplier:  Ampac™ 400F31MB
 View more versions of this product
Catalog No. 01-812-780
Explore available promotions
Description
Description
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
  • 3-ply laminated structure
  • Comprised of polyester, foil and polyethylene (PET/Foil/PE)
  • Offers oxygen and moisture barrier
  • Seal using Ampac's multi-purpose heat sealer
Order Info
Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.
Specifications
Specifications
900 Series
Foil
-56°C to +115°C
6 x 4 in. (15.2 x 10.2cm)
15.2 cm
10.2 cm
4 oz.
2400/Cs.
2.50 mil
6 in.
4 in.