Ampac™ Flexibles Silver Pak Bags
The 600 Series metalized pouches are ideal for powders and other granular materials
Supplier:  Ampac™ 604B1M
 View more versions of this product
Catalog No. 01-812-400
Explore available promotions
Description
Description
SilverPAK ofers a 2-ply laminated structure consisting of metalized polyester and polyethylene (METPET/PE).
Specifications
Specifications
2.50 mil
12 in.
8 in.
Product Suggestions
Product Suggestions