Eisco™ Amphibolite Specimen (Metamorphic Rock)
For use in studying hydrothermal alteration of rocks and minerals. EISCO Amphibolite Specimen (Metamorphic Rock) is formed from hornblende that has undergone significant amount of differential pressure.
Supplier:  Eisco™ ESNG0052
 View more versions of this product
Catalog No. S24774
Explore available promotions
Description
Description
  • Formed from further pressure and thermal alteration of hornblende, fusing and foliating mineral crystals
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Ideal for students studying hydrothermal alteration of rocks and minerals
  • Crystals within rock have been foliated, indicating direction of stress that rock experienced
  • Comes in clear labeled and barcoded bag for storage
Specifications
Specifications
Clear labeled and barcoded bag
Studying hydrothermal alteration of rocks and minerals
Approx. 1 in.
Each
Amphibolite Study Specimen (Metamorphic Rock)
Approx. 3 cm
Product Suggestions
Product Suggestions