missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Amphiregulin Human ELISA Kit DFS Item

Sandwich ELISA Kit

$794.00 - $794.00

Specifications

Accession Number P15514
Assay Sensitivity 10 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
EHAREG DFS Item
View Documents
Invitrogen™
EHAREG
96 Tests
Pack for $794.00
 
Description

Description

The Human Amphiregulin ELISA quantitates Hu Amphiregulin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Amphiregulin. Principle of the method The Human Amphiregulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Amphiregulin (AR) binds to EGF-Receptor (EGFR) with lower affinity than EGF. The mature secreted form AR is an 84-amino acid residue glycosylated polypeptide growth regulator, which is generated by proteolytic processing of a 252-amino acid transmembrane precursor. Seven different polypeptide ligands, which derive from distinct genes, are capable of binding to the extracellular domain of EGFR. These ligands include EGF, TGFalpha, AR, HB-EGF, cripto-1, epiregulin, and beta cellulin. All of these growth factors contain a characteristic EGF-like domain which is defined by six evenly spaced cysteine residues that generate three loops through the formation of disulfide bonds. AR protein has been localized by immunohistochemistry to the epithelium of the colon, stomach, pancreas, breast, and placenta. AR is reportedly overexpressed in human cancers of breast, colon, stomach, and pancreas.
Specifications

Specifications

P15514
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
ARB,CRDGF,SDGF,AREG
<10%
RUO
2°C to 8°C
Human
4 hr. 45 min.
10 pg/mL
Biotin
GCPR
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
374
<12%
HRP
Neurobiology
ARB, CRDGF, SDGF, AREG
1 hr. 20 min.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'