missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents
Supplier: Fisher BioReagents BP264520
Description
Amphotericin B is effective against yeast and fungi. An antibiotic solution used in cell biology.Specifications
Amphotericin B | |
250μg/mL | |
1397-89-3 | |
MFCD00877763 | |
Poly Bottle | |
C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O | |
924.09 | |
924.1 | |
Yellow | |
100°C |
Amphotericin B | |
Liquid | |
C47H73NO17 | |
15, 582 | |
APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
(1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car | |
134129663 | |
Filter Sterilized | |
20 mL |
Chemical Identifiers
1397-89-3 | |
924.09 | |
APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
(1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car |
C47H73NO17 | |
MFCD00877763 | |
134129663 | |
C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
Safety and Handling
Recommended Storage : -20°C