missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents

Supplier:  Fisher BioReagents BP264550

 View more versions of this product

Catalog No. BP264550



Explore available promotions

Description

Description

Amphotericin B is effective against yeast and fungi. An antibiotic solution used in cell biology.
Specifications

Specifications

Amphotericin B
250μg/mL
1397-89-3
MFCD00877763
Poly Bottle
C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
924.09
924.1
Yellow
100°C
Amphotericin B
Liquid
C47H73NO17
15, 582
APKFDSVGJQXUKY-ZNVUZQDLSA-N
(1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
134129663
Filter Sterilized
50 mL

Chemical Identifiers

1397-89-3
924.09
APKFDSVGJQXUKY-ZNVUZQDLSA-N
(1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
C47H73NO17
MFCD00877763
134129663
C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS Documents

Documents

Promotions

Promotions