missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Amprenavir

$195.98 - $580.84

Chemical Identifiers

CAS 161814-49-9
Molecular Formula C25H35N3O6S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466980250
View Documents
Thermo Scientific Chemicals
466980250
25 mg
Each for $195.98
 
AC466981000
View Documents
Thermo Scientific Chemicals
466981000
100 mg
Each for $580.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

161814-49-9
C25H35N3O6S

Specifications

161814-49-9
C25H35N3O6S
YMARZQAQMVYCKC-OEMFJLHTSA-N
(3S)-oxolan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
Amprenavir
25 mg
505.63
CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CCOC1)S(=O)(=O)C1=CC=C(N)C=C1
505.63
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only