missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amyl 4-Carboxyphenyl Carbonate 96.0+%, TCI America™
Supplier: TCI America C07721G
Specifications
Amyl 4-Carboxyphenyl Carbonate | |
141°C | |
C13H16O5 | |
1 g | |
RMIZZENBDILCRU-UHFFFAOYSA-N | |
4-{[(pentyloxy)carbonyl]oxy}benzoic acid | |
18352252 | |
≥96.0% (T) |
377085-56-8 | |
White | |
MFCD00059647 | |
4-Carboxyphenyl Amyl Carbonate, 4-Carboxyphenyl Pentyl Carbonate | |
CCCCCOC(=O)OC1=CC=C(C=C1)C(O)=O | |
252.27 | |
252.27 | |
Crystalline Powder |
Chemical Identifiers
377085-56-8 | |
252.27 | |
RMIZZENBDILCRU-UHFFFAOYSA-N | |
18352252 | |
CCCCCOC(=O)OC1=CC=C(C=C1)C(O)=O |
C13H16O5 | |
MFCD00059647 | |
4-Carboxyphenyl Amyl Carbonate, 4-Carboxyphenyl Pentyl Carbonate | |
4-{[(pentyloxy)carbonyl]oxy}benzoic acid |
Safety and Handling
TSCA : No