missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ AnaeroPack™ 7.0L Rectangular Jar

Mitsubishi™ AnaeroPack 7.0L Rectangular Jar is an anaerobic jar with easy-close lid.

Supplier:  Thermo Scientific™ RE685070

Catalog No. 23-246-387


Explore available promotions

Description

Description

Mitsubishi™ AnaeroPack 7.0L Rectangular Jar is an anaerobic jar with easy-close lid.

  • Holds 42 standard monoplates (12 x 85mm), 45 spacesaver plates (10 x 100mm), or six 150mm plates
Specifications

Specifications

AnaeroPack Rectangular Jar
Thermo Scientific Mitsubishi
7.0 L
Complete Box
Each
Videos
SDS
Documents

Documents

Promotions

Promotions