Thermo Scientific™ AnaeroPack™-MicroAero Gas Generator
Mitsubishi™ AnaeroPack-MicroAero is used for cultivation of microaerophilic microorganisms
Supplier:  Thermo Scientific™ RE681005
Catalog No. R681005
Explore available promotions
Description
Description
Mitsubishi™ AnaeroPack-MicroAero is used for cultivation of microaerophilic microorganisms such as Campylobacter jejuni and Helicobacter pylori without the addition of water.
  • Use 1 sachet/2.5L jar or 3 sachets/7.0L jar
Specifications
Specifications
AnaeroPack-MicroAero
Gas Generation Sachet
Thermo Scientific Mitsubishi
20/Pk.