Thermo Scientific™ AnaeroPouch™-MicroAero Gas Generator
Mitsubishi™ AnaeroPouch-MicroAero is used for cultivation of microaerophilic microorganisms w/o water.
Supplier:  Thermo Scientific™ RE682005
Catalog No. R682005
Explore available promotions
Description
Description
Mitsubishi™ AnaeroPouch-MicroAero Gas Generator is used for cultivation of microaerophilic microorganisms such as Campylobacter jejuni and Helicobacter pylori without the addition of water.
Use one sachet/AnaeroPouch-Bag or 0.4L jar with two plates
Specifications
Specifications
AnaeroPouch-MicroAero
Gas Generation Sachet
Thermo Scientific Mitsubishi
20/Pk.