missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Analytical Individual Weights: ASTM™ Class 3, With Calibration Certificate

Low magnetic permeability and high corrosion resistance for reliable stable values. Supplied with a calibration certificate.

$216.40 - $2097.13

Specifications

Certifications/Compliance With certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard
Classification ASTM Class 3
Material Stainless Steel
Density 7.9kg/dm3 at 20°C
Includes Plastic box
View More Specs

Products 13
Catalog Number Mfr. No. Mass Product Type Shape Price Quantity  
Catalog Number Mfr. No. Mass Product Type Shape Price Quantity  
01-915-802
View Documents
Mettler Toledo™
11123565
20 mg Analytical Sheet Weight Square
Each for $216.40
 
01-915-804
View Documents
Mettler Toledo™
11123567
100 mg Analytical Sheet Weight Square
Each for $216.40
 
01-915-805
View Documents
Mettler Toledo™
11123568
200 mg Analytical Sheet Weight Square
Each for $216.40
 
01-915-806
View Documents
Mettler Toledo™
11123569
500 mg Analytical Sheet Weight Square
Each for $216.40
 
01-915-807
View Documents
Mettler Toledo™
11123570
1 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $251.49
 
01-915-810
View Documents
Mettler Toledo™
11123573
10 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $265.14
 
01-915-811
View Documents
Mettler Toledo™
11123574
20 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $274.89
 
01-915-814
View Documents
Mettler Toledo™
11123576
100 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $299.26
 
01-915-816
View Documents
Mettler Toledo™
11123577
200 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $492.26
 
01-915-817
View Documents
Mettler Toledo™
11123578
500 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $594.62
 
01-915-818
View Documents
Mettler Toledo™
11123579
1 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $1,077.12
 
01-915-819
View Documents
Mettler Toledo™
11123580
2 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $1,584.00
 
01-915-820
View Documents
Mettler Toledo™
11123581
5 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $2,097.13
 
Description

Description

Features

  • Premium stainless steel
  • State-of-the-art turning procedures, improved coatings, stronger substrates
  • Electrolytic polishing ensures smooth, glossy surface
  • Magnetization and susceptibility of all weights are strictly controlled to ensure compliance with standards
Construction
  • Cylindrical knob weights and sheet weights; made of steel
  • Two-piece screw-knob construction for weights 1g and larger
  • Density: 7.9kg/dm3
Calibration
  • ISO/IEC 17025 accredited calibration laboratories
  • Lab cleans and adjusts weights and documents results in a calibration certificate
Includes: Weight, plastic box.

 

Specifications

Specifications

With certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard
Stainless Steel
Plastic box
1
ASTM Class 3
7.9kg/dm3 at 20°C
Each
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'