missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anatomical Chart Series - Pharynx and Larynx Anatomical chart

Highly detailed chart ideal for school use.

Supplier:  Customer Channels Group (Ccg) 9879DL

Catalog No. S9879PL


Explore available promotions

Description

Description

 

  • Printed on high-quality paper with flexible1.5 mil plastic lamination
  • Metal eyelets in top corners for wall hanging or for use with chart stands
  • Write on and wipe-off
  • Easily rolls up
  • Measures 20 x 26 in.

 

Specifications

Specifications

Science
Laminated
20 in.
Pharynx/Larynx Poster
26 in.
Metal eyelets in top corners for wall hanging or for use with chart stands
Videos
SDS
Documents

Documents

Promotions

Promotions