Andwin Scientific 100-Slide Capacity Microscope Slide Storage Boxes
Designed for safe, long-term slide storage
Supplier: Andwin Scientific AND880110
Description
- Constructed from polystyrene
- 100-Slide capacity
- Corrosion-resistant hinges and clasp
- Cork-lined bottoms
- Individually numbered slide positions
- Available in four colors
Specifications
100 | |
100 | |
Cork lined bottom |
Blue | |
Microscope Slide | |
Polystyrene |