missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anhydrous Lactose, NF, Spectrum™ Chemical
LA103, 63-42-3, C12H22O11
Supplier: Spectrum Chemical Mfg Cor LA10325KG
Description
Spectrum™ Chemical Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
63-42-3 | |
0.1% | |
Poly Pail | |
2.5 kg | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
NF |
100% | |
0.5% | |
C12H22O11 | |
+54.4° to +55.9° | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
342.30 |
Chemical Identifiers
63-42-3 | |
342.30 | |
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
C12H22O11 | |
GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |