missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aniline-2-sulfonic Acid, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 400640250

 View more versions of this product

Catalog No. AC400640250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aniline-2-sulfonic acid
>300°C
Authentic
Glass bottle
H2NC6H4SO3H
25 g
15, 6978
Solubility in water: 3100mg/l (23°C)
C1=CC=C(C(=C1)N)S(=O)(=O)O
173.19
CHEBI:1015
95%
88-21-1
White to Beige
95%
C6H7NO3S
MFCD00007705
14, 681
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
2-aminobenzenesulfonic acid
6926
173.19
Crystalline Powder

Chemical Identifiers

88-21-1
173.19
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
6926
2-aminobenzenesulfonic acid
C6H7NO3S
MFCD00007705
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
CHEBI:1015
C1=CC=C(C(=C1)N)S(=O)(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 201-810-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only