missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99+%, ACS reagent

CAS: 62-53-3 | C6H7N | 93.13 g/mol

$62.66 - $725.15

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423420050
SDS View Documents
Thermo Scientific Chemicals
423420050
5 mL Glass bottle
Each for $62.66
Only null left
 
AC423425000
SDS View Documents
Thermo Scientific Chemicals
423425000
500 mL Glass Bottle
Each for $182.05
Case of 6 Each for $0.00
Only null left
 
AC423420010
SDS View Documents
Thermo Scientific Chemicals
423420010
1 L Glass Bottle
Each for $310.29
Only null left
 
AC423420025
SDS View Documents
Thermo Scientific Chemicals
423420025
2.5 L Glass bottle
Each for $725.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62-53-3
93.13
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
6115
aniline
C6H7N
MFCD00007629
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
CHEBI:17296
NC1=CC=CC=C1

Specifications

62-53-3
Colorless to White
181.0°C to 185.0°C
0.005 % max.
99% min. (GC)
C6H7N
C6H5NH2
5 mL
06,21; 17,16
15, 652
Solubility in water: 36g/L (20°C).
NC1=CC=CC=C1
93.13
4.4 mPa.s (20°C)
93.13
99+%
Crystalline Powder or Crystals
-6.2°C
1.0213g/mL
76°C
Authentic
Glass bottle
1.5845 to 1.5865
MFCD00007629
12, 59
1.0213
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
aniline
6115
CHEBI:17296
0.5mmHg at 20°C
ACS Reagent
Aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 200-539-3

Recommended Storage : this product darkens to reddish-brown on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only