missing translation for 'onlineSavingsMsg'
Learn More

Aniline Blue, Water Soluble, Spectrum™ Chemical

$303.83 - $4563.93

Chemical Identifiers

CAS 28631-66-5
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608867
View Documents
Spectrum Chemical Mfg Cor
AN10510GM
10 g Amber Glass Bottle
Each for $303.83
Only null left
 
18608869
View Documents
Spectrum Chemical Mfg Cor
AN10525GM
25 g Amber Glass Bottle
Each for $545.20
Only null left
 
18608870
View Documents
Spectrum Chemical Mfg Cor
AN105100GM
100 g Amber Glass Bottle
Each for $1,085.03
Only null left
 
18608868
View Documents
Spectrum Chemical Mfg Cor
AN1051KGBL
1 kg Amber Glass Bottle
Each for $4,563.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

28631-66-5

Specifications

28631-66-5
Reagent
Dark Red
Aniline Blue
1
Amber Glass Bottle
10 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.