missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aniline hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1302436

 View more versions of this product

Catalog No. AAA1302436


Explore available promotions

Description

Description

Aniline hydrochloride was used in the preparation of polyaniline coated poly(styrene-co-styrene sulfonate) nanoparticles. It was used to study the induction of Nei-like DNA glycosylases (NEIL1/2)-mediated base excision repair(BER) in rat spleen and 8-oxoguanine glycosylase 1-mediated BER due to aniline exposure. It is used in printing inks, clothing marking inks, paints and paint removers. It is also used as a stove polisher and shoe polisher, as well as in the synthesis of dyes, crayons and antioxidants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline hydrochloride was used in the preparation of polyaniline coated poly(styrene-co-styrene sulfonate) nanoparticles. It was used to study the induction of Nei-like DNA glycosylases (NEIL1/2)-mediated base excision repair(BER) in rat spleen and 8-oxoguanine glycosylase 1-mediated BER due to aniline exposure. It is used in printing inks, clothing marking inks, paints and paint removers. It is also used as a stove polisher and shoe polisher, as well as in the synthesis of dyes, crayons and antioxidants.

Solubility
Soluble in water (1070 g/L).

Notes
It is light sensitive and hygroscopic. Incompatible with oxidizing agents.
Specifications

Specifications

Aniline hydrochloride
196°C to 200°C
245°C
C6H8ClN
500 g
3593823
14,659
Soluble in water (1070g/L).
[H+].[Cl-].NC1=CC=CC=C1
129.59
129.59
142-04-1
1.221
193°C (379°F)
MFCD00012958
UN1548
Hygroscopic; Light sensitive
aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline
MMCPOSDMTGQNKG-UHFFFAOYSA-N
aniline;hydrochloride
8870
99%

Chemical Identifiers

142-04-1
129.59
MMCPOSDMTGQNKG-UHFFFAOYSA-N
8870
C6H8ClN
MFCD00012958
aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline
[H+].[Cl-].NC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H318-H341-H351-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANILINE HYDROCHLORIDE

EINECSNumber : 205-519-8

RTECSNumber : CY0875000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only