Learn More
Aniline hydrochloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158411000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aniline hydrochloride | |
142-04-1 | |
Cream-Yellow to White | |
193°C | |
Authentic | |
Plastic bottle | |
C6H5NH2·HCl | |
100 g | |
aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline | |
MMCPOSDMTGQNKG-UHFFFAOYSA-N | |
aniline;hydrochloride | |
8870 | |
99% |
99% | |
196.0°C to 199.0°C | |
245.0°C | |
0.5 % max. | |
99% | |
C6H8ClN | |
MFCD00012958 | |
15,652 | |
Solubility in water: 1070g/L (25°C). Other solubilities: insoluble in ethyl ether and chloroform | |
[H+].[Cl-].NC1=CC=CC=C1 | |
129.59 | |
129.59 | |
Powder |
Chemical Identifiers
142-04-1 | |
129.59 | |
MMCPOSDMTGQNKG-UHFFFAOYSA-N | |
8870 | |
[H+].[Cl-].NC1=CC=CC=C1 |
C6H8ClN | |
MFCD00012958 | |
aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline | |
aniline;hydrochloride |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Suspected of causing genetic defects.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Danger
EINECSNumber : 205-519-8
RTECSNumber : CY0875000
TSCA : TSCA
RUO – Research Use Only