Stoelting™ Animal Markers
Animal Markers allow for easy, long-lasting identification of laboratory animals.
Supplier: Stoelting™ 504491
Description
- Nontoxic and devoid of any hazardous materials
- A single marker can be used to mark 300 to 500 rats (depending upon animal size and/or applied area)
- Once applied, color remains on fur for 6 to 12 weeks; does not work on bare skin
- Broad and fine tip markers available
Specifications
Neuroscience | |
Rodent | |
Single Packaging |
Fine tip | |
Orange |