missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anisoin 97.0+%, TCI America™
$118.51 - $339.70
Chemical Identifiers
CAS | 119-52-8 |
---|---|
Molecular Formula | C16H16O4 |
Molecular Weight (g/mol) | 272.30 |
MDL Number | MFCD00008411 |
InChI Key | LRRQSCPPOIUNGX-UHFFFAOYNA-N |
Synonym | anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one |
PubChem CID | 95415 |
IUPAC Name | 2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one |
SMILES | COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 |
Chemical Identifiers
119-52-8 | |
272.30 | |
LRRQSCPPOIUNGX-UHFFFAOYNA-N | |
95415 | |
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 |
C16H16O4 | |
MFCD00008411 | |
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one | |
2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one |
Specifications
119-52-8 | |
White-Yellow | |
MFCD00008411 | |
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one | |
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 | |
272.30 | |
272.30 | |
Crystalline Powder |
112°C | |
C16H16O4 | |
25 g | |
LRRQSCPPOIUNGX-UHFFFAOYNA-N | |
2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one | |
95415 | |
≥97.0% (HPLC) | |
Anisoin |
Safety and Handling
EINECSNumber : (4)-0277
TSCA : Yes