missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ Annexin V Conjugates for Apoptosis Detection

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.

$520.00 - $672.00

Specifications

Product Type Annexin V conjugate
Shipping Condition Wet Ice
For Use With (Equipment) Flow Cytometer
View More Specs

Products 9
Catalog Number Mfr. No. Conjugate Excitation/Emission Flow Cytometer Laser Lines Price Quantity  
Catalog Number Mfr. No. Conjugate Excitation/Emission Flow Cytometer Laser Lines Price Quantity  
A13201 DFS Item
View Documents
Molecular Probes™
A13201
Alexa Fluor 488 495/519 488 N/A
 
A13199 DFS Item
View Documents
Molecular Probes™
A13199
FITC 494/518 488 N/A
 
501121497 DFS Item
View Documents
Molecular Probes™
A13202
Alexa Fluor 568 578/603 532, 561 N/A
 
A13203 DFS Item
View Documents
Molecular Probes™
A13203
Alexa Fluor 594 590/617 532 N/A
 
501121498 DFS Item
View Documents
Molecular Probes™
A13204
Biotin-X N/A
 
A23204 DFS Item
View Documents
Molecular Probes™
A23204
Alexa Fluor 647 650/665 633-637 N/A
 
A35108
View Documents
Molecular Probes™
A35108
Alexa Fluor 555 555/565 532, 561 N/A
 
501121504 DFS Item
View Documents
Molecular Probes™
A35109
Alexa Fluor 680 679/702 633-637 N/A
 
501121505 DFS Item
View Documents
Molecular Probes™
A35122
Pacific Blue 410/455 405 N/A
 
Description

Description

Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.

Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

Specifications

Specifications

Annexin V conjugate
Wet Ice
Flow Cytometer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'