Learn More
Ansell™ Chemical- and Flame-Resistant Bib Pants
Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration
$568.13 - $568.13
Specifications
| Suspender | Elastic |
|---|---|
| Construction | Trilaminate |
| For Use With (Application) | Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals |
| Length (English) Inseam | 31 in. |
| Length (Metric) Inseam | 79 cm |
Alert:
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.
| Catalog Number | Mfr. No. | Color | Material | Size | Size (English) Waist | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Material | Size | Size (English) Waist | Price | Quantity | ||
|
17-987-449D
|
Ansell™
66662L |
Red | CPC Polyester | Large | 52 in. |
Each for $568.13
Case of 5 Each for $2,840.65
|
|
||
Description
Gore membrane is a bicomponent PTFE membrane that prevents liquid penetration but allows a vapor permeation, allowing perspiration to evaporate and cool the wearer.
- Garments stitched with Gore thread
- All seams are heat and pressure sealed with Gore-Seam™ tape to prevent liquid penetration
- Fabrics are durable, flexible, tear and snag resistant, washable, and reusable
Coveralls
- Feature two-way, heavy-duty, static-free zipper
- Double storm flap; stand-up collar; raglan sleeves
- Hook-and-loop closures on storm flap, collar, sleeves, and leg bottoms
- Heavy-duty zipper
- Double storm flap; stand-up collar; raglan sleeves
- Hook-and-loop closures on storm flap, collar, and sleeves
- Sized to be worn over clothing
- Elastic suspenders with quick-release clips for fast removal
- Leg bottoms have hook-and-loop closures
- Attach, by means of a hook-and-loop fastener, to back of collar on coveralls or jacket
- Drawstring closure gives snug fit over hardhat
Order Info
For chemical splash protection, choose CPC garments made with red polyester Gore ChemSplash fabric (7 oz. per sq. yd.). When flame protection is also required, choose garments made with blue aramid (Nomex™) Gore ChemSplash fabric (6 oz. per sq. yd.); when exposed to flame, it will self-extinguish when removed from the flame source, and not contribute to burn injury.
Specifications
| Elastic | |
| Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals | |
| 79 cm |
| Trilaminate | |
| 31 in. |