Learn More
Ansell™ Chemical- and Flame-Resistant Jackets
Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration
Supplier: Ansell™ 66660L
Alert:
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.
Description
Gore membrane is a bicomponent PTFE membrane that prevents liquid penetration but allows a vapor permeation, allowing perspiration to evaporate and cool the wearer.
- Garments stitched with Gore thread
- All seams are heat and pressure sealed with Gore-Seam™ tape to prevent liquid penetration
- Fabrics are durable, flexible, tear and snag resistant, washable, and reusable
Jackets
- Heavy-duty zipper
- Double storm flap; stand-up collar; raglan sleeves
- Hook-and-loop closures on storm flap, collar, and sleeves
- Sized to be worn over clothing
Order Info
For chemical splash protection, choose CPC garments made with red polyester Gore ChemSplash fabric (7 oz. per sq. yd.). When flame protection is also required, choose garments made with blue aramid (Nomex™) Gore ChemSplash fabric (6 oz. per sq. yd.); when exposed to flame, it will self-extinguish when removed from the flame source, and not contribute to burn injury.
Specifications
Large | |
Red | |
NFPA | |
Stand-up | |
137.1cm | |
30 in. | |
76.2cm |
CPC Polyester | |
Double Hook and Loop Front Flap | |
Trilaminate | |
54 in. | |
Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals | |
34 in. | |
86.3cm |