Ansell™ Polyethylene Aprons
Disposable, provide protection for light- and medium-duty applications
Supplier:  Ansell™ 5429011
Catalog No. 19-037-892
Explore available promotions
Description
Description
High-quality virgin material combines flexibility and resistance to chemicals, fats and oils.
  • Easy to clean
  • Embossed
  • Conveniently packaged
  • 600/Cs.
Food preparation, processing and handling; food service/ catering; janitorial and maintenance work
Compliance
Complies with FDA regulations for food contact.
Specifications
Specifications
Polyethylene
Single-use
1.75 mil
White
Embossed
71.1 x 114.3 cm. (28 x 45 in.)
Product Suggestions
Product Suggestions