missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell AlphaTec™ Solvex™ 37-145 Nitrile Gloves

Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.

Supplier:  Ansell 3714511060

 View more versions of this product

Catalog No. 11-391-136A



Explore available promotions

Description

Description

Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.

  • Nitrile construction protects against abrasions, cuts, snags and punctures
  • Curved finger design and contoured palm help reduce fatigue
  • Full cut for easy donning and doffing
  • Dark Green
  • No inner cotton flocking, reducing risk of lint contamination
  • Sandpatch finish provides smooth surface
  • Fully reusable, comfortable
  • Complies with FDA food handling requirements
Specifications

Specifications

ANSI 5
Yes
ANSI A4
Nitrile
11 mil
EN 388:4001, EN 388:3001X
Type A
Bacteria, Fungi only
Gauntlet
Unlined
13 in.
FDA 21 CFR 177.2600
Green
Yes
6
0.27 mm
No
JKNOPT
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Sandpatch
37-145 6 117071
33 cm
SDS
Documents

Documents

Promotions

Promotions