missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansul™ Bulk SPILL-X™ Agents
For response to larger spills
$2191.97 - $2191.97
Specifications
Capacity (English) | 5 gal. |
---|---|
Capacity (Metric) | 19 L |
For Use With (Equipment) | Spill Gun™ applicator |
Includes | Detailed instructions for responding to a spill |
Alert:
Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste material should comply with local, state, and federal regulations.
Catalog Number | Mfr. No. | Base | Color | Weight (English) | Weight (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Base | Color | Weight (English) | Weight (Metric) | Price | Quantity | |||
17-200-1A
|
Ansul™
76255 |
Magnesium Oxide | Red | 50 lb. | 22.7 kg |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
17-200-1C
|
Ansul™
76396 |
Citric Acid | Tan | 42 lb. | 19 kg |
Each for $2,191.97
|
|
|||
17-978
|
Ansul™
78443 |
Urea | Green | 37 lb. | 16.8 kg |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
- Use a scoop or shovel to apply the agent directly onto the spill
- Bulk-packed agents can also be used to refill a Spill-Gun™ applicator (not included)
- Include detailed instructions for responding to a spill
Specifications
5 gal. | |
19 L | |
Spill Gun™ applicator | |
Detailed instructions for responding to a spill |