Ansul™ Treatment Refills Kit for Chemical Spill Kits
Minimize downtime - have refills ready at hand
Supplier:  Ansul™ 78919
Catalog No. 17-987-144F
Explore available promotions
Includes: Safety goggles, rubber gloves, disposal bags, scraper, pick-up pans, pH test papers, and spill treatment guide
Description
Description
General refills for spill response kits. Replacement SPILL-X™ agents sold separately.
Specifications
Specifications
Refill Spill Kit
Goggles, rubber gloves, bags, scraper, pick-up pans, pH test paper, and spill treatment guide