missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anthracene-9-carboxylic acid, 98+%, Thermo Scientific Chemicals

$132.36 - $1761.38

Chemical Identifiers

CAS 723-62-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00001257
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
PubChem CID 2201
ChEBI CHEBI:34507
IUPAC Name anthracene-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1404909
View Documents
Thermo Scientific Chemicals
A1404909
10 g
Each for $132.36
 
AAA1404918
View Documents
Thermo Scientific Chemicals
A1404918
50 g
Each for $474.63
 
AAA1404930
View Documents
Thermo Scientific Chemicals
A1404930
250 g
Each for $1,761.38
 
Description

Description

Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
Specifications

Chemical Identifiers

723-62-6
222.24
XGWFJBFNAQHLEF-UHFFFAOYSA-N
2201
anthracene-9-carboxylic acid
C15H10O2
MFCD00001257
9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
CHEBI:34507
OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

723-62-6
C15H10O2
10 g
9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
XGWFJBFNAQHLEF-UHFFFAOYSA-N
anthracene-9-carboxylic acid
2201
222.24
Anthracene-9-carboxylic acid
∼218°C (decomposition)
MFCD00001257
1875336
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene,chloroform,and hydronaphthalenes. Insoluble in water.
OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
222.24
CHEBI:34507
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 211-964-9

RTECSNumber : CB8764000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only