Learn More
Bax Mouse anti-Human, Clone: 2D2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB4601
Description
Anti-Bax Antibody, N-terminus, aa3-16 hBax protein, clone 2D2 is an antibody against Bax for use in immunofluoresence, western blot, immunohistochemistry (paraffin).Specifications
| Bax | |
| Monoclonal | |
| Unconjugated | |
| NM_004324.3; NM_138761.2; NM_138763.2; NM_138764.2; NM_138765.2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Purified |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 2D2 | |
| Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide. | |
| BAX | |
| Synthetic peptide corresponding to aa 3-16 (GSGEQPRGGGPTSS) of human bax. This amino acid sequence is not shared by mouse or rat Bax. | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. | |
| IgG1 |
For Research Use Only