missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1 Mouse anti-Human, Hamster, Clone: JSB-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB4120
Description
Anti-MDR1 Antibody, clone JSB-1 detects level of MDR1 and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry (paraffin) and western blotting.Specifications
| MDR1 | |
| Monoclonal | |
| Unconjugated | |
| NM_000927.3 | |
| Mouse | |
| 100 μg | |
| Hamster, Human | |
| Supernatant |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| JSB-1 | |
| UnPurified mouse culture supernatant containing 0.7% BSA and 0.1% sodium azide as a preservative. | |
| ABCB1, PGY1, MGC163296, GP170, ABC20, CD243, P-gp, MDR1, CLCS, EC 3.6.3.44 | |
| Unpurified | |
| Primary | |
| −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
For Research Use Only