missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva
Highly species-specific anti-Mouse IgG. Cytiva Lifescience™ anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody is designed for applications such as protein blotting, ELISA and immunocytochemistry.
$134.05 - $505.88
Specifications
Antigen | IgG, Whole Molecule |
---|---|
Classification | Monoclonal |
Conjugate | HRP |
Host Species | Sheep |
Isotype | IgG |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
45-001-275
|
Cytiva
NA931100UL |
0.1 mL |
Each for $134.05
|
|
|||||
45000679
|
Cytiva
NA9311ML |
1 mL |
Each for $505.88
|
|
|||||
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Specifications
IgG, Whole Molecule | |
HRP | |
IgG | |
Affinity Purified |
Monoclonal | |
Sheep | |
Secondary |