Sartorius Anti-Static Devices for Balances
Control charges on samples
Supplier: Sartorius YIB010UR
Description
- Anti-static pen—direct the pen at item to be neutralized
- Ionizing blower—point blower toward sample prior to weighing
Specifications
CPA Balance, Extended Balance, SE and ME Ultramicro Balance | |
8 cm | |
12 cm |
3.1 in. | |
4.7 in. | |
Ionizing Blower |