Sartorius Anti-Static Devices for Balances
Control charges on samples
Supplier:  Sartorius YSTP01
 View more versions of this product
Catalog No. 14-557-540
Explore available promotions
Description
Description
  • Anti-static pen—direct the pen at item to be neutralized
  • Ionizing blower—point blower toward sample prior to weighing
Specifications
Specifications
CPA Balance, Extended Balance, SE and ME Ultramicro Balance
Anti-static Pen