Innovatek Anti-Sterptolysin O (ASO) Latex Test Kits
Innovatek Anti-Sterptolysin O (ASO) Latex Test Kits offer results in less than 3 minutes
Supplier:  Innovatek 419110003
 View more versions of this product
Catalog No. 29-801-064
Explore available promotions
Description
Description
  • Fast 2 to 3 minute rotation
  • Complete kit includes all reagents and controls
  • Disposable test cards
  • 6 to 12 months of shelf life
  • Sensitive and specific
Specifications
Specifications
Anti-Streptolysin O (ASO)
Single-use
100 Tests/Kit
≈3 min.
Product Suggestions
Product Suggestions