Molecular Probes™ Antibody Conjugate Purification Kit, 50-100 μg conjugate DFS Item
Antibody Conjugate Purification Kit, 50-100 μg conjugate
Supplier:  Molecular Probes™ A33088
Catalog No. A33088
Explore available promotions
Description
Description
Molecular Probes™ Antibody Conjugate Purification kits efficiently purify antibody conjugates from unconjugated dye or biotin. This particular kit is optimized for the purification of 50–100 μg of antibody conjugate. Purification is accomplished using an easy-to-assemble size-exclusion spin column. Each kit contains sufficient material for 5 separate purifications, and can also be used to purify high-molecular weight proteins (> 30 kDa) away from unconjugated labels.
Additional Features of this Antibody Conjugate Purification Kit:
• Purified conjugate ready-to-use in ∼45 minutes with ∼15 minutes of hands-on time
• Designed for low-molecular weight labels (less than 3000 Da, e.g. Alexa Fluor™ dyes); cannot be used for removing unconjugated high-molecular weight labels (>10 kDa) such as enzymes (HRP) or proteins (R-phycoerythrin)
More about Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental set-up. To learn more about our labeling kits, see Kits for Labeling Proteins and Nucleic Acids in The Molecular Probes™ Handbook or visit our antibody labeling page.
Specifications
Specifications
Antibody Conjugate Purification Kit
5 purifications
Store in refrigerator (2-8 °C).
Spin Column
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.