missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony, 99+%, powder, 0.15 mm, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 192450100

Catalog No. AC192450100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
p.a., 0.15 mm
7440-36-0
6.68
Sb
10 g
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
121.75
Glass bottle
Antimony
630°C
1635°C
99% min. (Iodometry)
MFCD00011211 MFCD00134030
15, 683
Solubility in water: insoluble
[Sb]
121.76
CHEBI:30304
≥99%
Powder

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer if inhaled.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 231-146-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only