missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis), Thermo Scientific Chemicals

Form: Broken pieces from cast rod

$362.70 - $1221.18

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4504736
View Documents
Thermo Scientific Chemicals
04504736
500 g
Each for $362.70
 
AA45047A3
View Documents
Thermo Scientific Chemicals
045047A3
2 kg
Each for $1,221.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
[Sb]
Sb
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304

Specifications

Silver to Gray
Silver Gray rod
Sb
500 g
WATWJIUSRGPENY-UHFFFAOYSA-N
distibyne
5354495
Odorless
Antimony
7440-36-0
99.8% (Metals basis)
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
Pieces
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-146-5

RTECSNumber : CC4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only