Learn More
Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
$72.30 - $677.51
Chemical Identifiers
CAS | 10025-91-9 |
---|---|
Molecular Formula | Cl3Sb |
Molecular Weight (g/mol) | 228.11 |
MDL Number | MFCD00011212 |
InChI Key | FAPDDOBMIUGHIN-UHFFFAOYSA-K |
Synonym | antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony |
PubChem CID | 24814 |
ChEBI | CHEBI:74856 |
SMILES | [Cl-].[Cl-].[Cl-].[Sb+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1194422
|
Thermo Scientific Chemicals
A1194422 |
100 g |
Each for $72.30
|
|
||||||
AAA1194436
|
Thermo Scientific Chemicals
A1194436 |
500 g |
Each for $189.92
|
|
||||||
AAA119440E
|
Thermo Scientific Chemicals
A119440E |
2500 g |
Each for $677.51
|
|
||||||
Description
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.
Solubility
Soluble in water, hydrochloric acid, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone and carbon tetrachloride. Insoluble in organic bases.
Notes
Moisture sensitive and hygroscopic in nature. It is corrosive to metals and tissue. Incompatible with strong oxidizing agents, strong acids and strong bases. Decomposes in water.
Chemical Identifiers
10025-91-9 | |
228.11 | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
24814 | |
[Cl-].[Cl-].[Cl-].[Sb+3] |
Cl3Sb | |
MFCD00011212 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
CHEBI:74856 |
Specifications
223.5°C | |
10025-91-9 | |
Cl3Sb | |
UN1733 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
trichlorostibane | |
24814 | |
228.11 | |
Pungent | |
3.14 |
73.4°C | |
100 g | |
MFCD00011212 | |
14,707 | |
Soluble in water,hydrochloric acid,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone and carbon tetrachloride. Insoluble in organic bases. | |
[Cl-].[Cl-].[Cl-].[Sb+3] | |
228.11 | |
CHEBI:74856 | |
≥99% | |
Moisture sensitive | |
Antimony (III) chloride |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY TRICHLORIDE, SOLID
EINECSNumber : 233-047-2
RTECSNumber : CC4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only