Learn More
Antimony potassium tartrate hydrate, 98%
CAS: 331753-56-1 | C8H4K2O12Sb2 | 613.83 g/mol
$92.85 - $1082.14
Chemical Identifiers
| CAS | 331753-56-1 |
|---|---|
| Molecular Formula | C8H4K2O12Sb2 |
| Molecular Weight (g/mol) | 613.83 |
| MDL Number | MFCD00148863 |
| InChI Key | GUJUCWZGYWASLH-UHFFFAOYNA-J |
| Synonym | potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate |
| IUPAC Name | dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1â´,â·]tetradecane-3,9-dicarboxylate |
| SMILES | [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1088922
|
Thermo Scientific Chemicals
A1088922 |
100 g |
Each for $92.85
|
|
|||||
|
AAA1088936
|
Thermo Scientific Chemicals
A1088936 |
500 g |
Each for $264.79
|
|
|||||
|
AAA108890E
|
Thermo Scientific Chemicals
A108890E |
2500 g |
Each for $1,082.14
|
|
|||||
Description
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
Chemical Identifiers
| 331753-56-1 | |
| 613.83 | |
| GUJUCWZGYWASLH-UHFFFAOYNA-J | |
| dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1â´,â·]tetradecane-3,9-dicarboxylate |
| C8H4K2O12Sb2 | |
| MFCD00148863 | |
| potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate | |
| [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2 |
Specifications
| 331753-56-1 | |
| 98% | |
| MFCD00148863 | |
| UN1551 | |
| Soluble in water. Insoluble in ethanol. | |
| [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2 | |
| 613.83 | |
| 98% |
| 2.607 g/mL | |
| C8H4K2O12Sb2 | |
| 100 g | |
| potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate | |
| GUJUCWZGYWASLH-UHFFFAOYNA-J | |
| dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1â´,â·]tetradecane-3,9-dicarboxylate | |
| 613.82 (anhydrous) | |
| Antimony potassium tartrate hydrate |
Safety and Handling
GHS H Statement
H302-H332
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POTASSIUM TARTRATE
EINECSNumber : 608-190-2
RTECSNumber : CC7350000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only