missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01010030
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Antimony powder | |
1380°C | |
Sb | |
UN2871 | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
[Sb] | |
121.76 | |
CHEBI:30304 | |
−100 Mesh | |
Apparent density: 2.4g/cm3 to 2.9g/cm3 |
7440-36-0 | |
(metals basis) | |
MFCD00011211 MFCD00134030 | |
250 g | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
antimony | |
5354495 | |
99.5% | |
Odorless | |
Powder |
Chemical Identifiers
7440-36-0 | |
121.76 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
5354495 | |
antimony |
Sb | |
MFCD00011211 MFCD00134030 | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
CHEBI:30304 | |
[Sb] |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
H335-H351-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POWDER
EINECSNumber : 231-146-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only