missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Antipain, MP Biomedicals™
$267.55 - $957.71
Chemical Identifiers
CAS | 37691-11-5 |
---|---|
Molecular Formula | C27H44N10O6 |
Molecular Weight (g/mol) | 604.71 |
MDL Number | MFCD00135959 |
InChI Key | SDNYTAYICBFYFH-TUFLPTIASA-N |
Synonym | antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid |
PubChem CID | 133126507 |
IUPAC Name | 2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid |
SMILES | CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15284305
|
MP Biomedicals Inc
0215284305 |
5 mg |
N/A
|
|
|||||
ICN15284325
|
MP Biomedicals Inc
0215284325 |
25 mg |
N/A
|
|
|||||
Chemical Identifiers
37691-11-5 | |
604.71 | |
SDNYTAYICBFYFH-TUFLPTIASA-N | |
133126507 | |
CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O |
C27H44N10O6 | |
MFCD00135959 | |
antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid | |
2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid |
Specifications
37691-11-5 | |
C27H44N10O6 | |
5 mg | |
SDNYTAYICBFYFH-TUFLPTIASA-N | |
2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid | |
133126507 | |
Powder |
Yellow | |
MFCD00135959 | |
antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid | |
CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O | |
604.71 | |
604.7 | |
Antipain |