missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Anza™ T4 DNA Ligase Master Mix
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
$383.00 - $1284.00
Specifications
Concentration | 4X |
---|---|
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Enzyme | T4 DNA Ligase |
Compatible Buffer | Elution Buffer, 1X Anza Buffer |
Product Type | T4 DNA Ligase Master Mix |
Includes: Anza™ T4 DNA Ligase Master Mix
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.- One Ligase for both cohesive and blunt end ligations
- 4X conc. enables higher volume of dilute DNA in reaction mixture
- Ligation complete in 15 minutes
- Convenient Master Mix format reduces pipetting
Certifications
CoA
Specifications
4X | |
T4 DNA Ligase | |
T4 DNA Ligase Master Mix |
Approved for shipment on Wet or Dry Ice | |
Elution Buffer, 1X Anza Buffer | |
Anza™ |
For Research Use Only. Not for use in diagnostic procedures.