M&I Materials Apiezon™ M Grease
Low vapor pressure sealing agents and lubricants for high-vacuum applications
Supplier: M&I Materials FAPGM00100
Description
Apiezon products are silicone-free, high-purity hydrocarbons distilled from selected petroleum feedstock in a molecular still.
- M Grease (14-638-15C) is inexpensive enough to be used as general-purpose grease
Specifications
M Grease | |
1.7 x 109 torr | |
50°F to 86°F | |
3.5 oz. |
Low vapor pressure sealing agents and lubricants for high-vacuum applications | |
5 Pk. | |
10°C to 30°C |