missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I or Buffer II DFS Item

AmpliTaq DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli.

Supplier:  Applied Biosystems™ N8080160

 View more versions of this product

Catalog No. N8080160


Explore available promotions

Description

Description

AmpliTaq DNA Polymerase is a 94-kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for the PCR process, and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial and lot-to-lot.

AmpliTaq DNA Polymerase is available with GeneAmp10X PCR Buffer I or with GeneAmp 10X PCR Buffer II and MgCl2 Solution.

Reliable and robust PCR

The thermal activity profile of AmpliTaq DNA Polymerase is good for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications.

Features

  • The most thoroughly characterized enzyme available for the PCR process
  • Excellent thermal activity profile makes it good for PCR applications
  • QC-tested to guarantee reproducible results

Notes

  • For superior PCR performance, we recommend DreamTaq DNA Polymerase.
Specifications

Specifications

• AmpliTaq DNA Polymerase (5 U/μL), 50 μL
• GeneAmp 10X PCR Buffer, 1.5 mL

Store at -20°C.
AmpliTaq DNA Polymerase
5' - 3'
250 Units
Standard PCR
1X
200 Reactions
Separate Components
DNA
Stand-alone enzyme
Standard
DNA Polymerase
Dry Ice
5 U/μL
No
3'-A
5 kb or less
Buffer I
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.