missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL DFS Item

Reduces PCR reaction time from 2 hours to as little as 25 minutes

Supplier:  Applied Biosystems™ 4346906

 View more versions of this product

Catalog No. 4346906


Explore available promotions

Description

Description

  • Integral component of Applied Biosystems Fast PCR System
  • Maximum thermal conductivity for precise thermal cycling
  • Reproducible, specific, and sensitive PCR results in just 25 minutes
  • Validated with other system components for consistent results
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Increased thermal contact for faster, more uniform heating
  • Advanced design dramatically increases its thermal contact with Applied Biosystems Veriti™ 96-Well Fast Thermal Cycler
  • Consistent well-to-well thickness helps all samples cycle through PCR process uniformly

  • Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System

Applications: Fast Real-Time PCR, PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Specifications

Includes Barcode
Contains 20 plates.

May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
MicroAmp
Microplate
Semi
0.2 mL
0.1 mL
Fast
Certified DNA/RNase Free
20 Plates
Optical
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polypropylene
96
20 Plates
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions