Applied Biosystems™ MicroAmp™ Optical 384-Well Reaction Plate with Barcode DFS Item
Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification
Supplier:  Applied Biosystems™ 4326270
 View more versions of this product
Catalog No. 4326270
Explore available promotions
Description
Description
  • Constructed from a single rigid piece of polypropylene in a 384-well format
  • Designed to work with Applied Biosystems 384-well Real-Time PCR systems and thermal cyclers
  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Platform: 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7900HT System, GeneAmp™ 9700, QuantSt
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
Specifications
Includes Barcode
Contains 10 packages of 50 plates each (500 plates).
May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
MicroAmp
Microplate
Skirted
40 μL
500 plates
Fast or Standard
Certified DNA/RNase Free
500 Plates
Optical
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polypropylene
384
500 plates