Applied Biosystems™ RapidHIT™ ID System
The Applied Biosystems RapidHIT ID System is a fast and simple-to-use instrument that produces trusted lab-quality forensic DNA profiles in as little as 90 minutes.
Supplier:  Applied Biosystems A41810
Catalog No. A41810
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The Applied Biosystems RapidHIT ID System is a fast and simple-to-use instrument that produces trusted lab-quality forensic DNA profiles in as little as 90 minutes. The system integrates sample preparation and capillary electrophoresis to generate DNA profiles that are aggregated within Applied Biosystems RapidLINK Software for direct upload to the database of choice or for further review and analysis.
The RapidHIT ID System delivers the reliable, reproducible, high-quality data you have come to expect from the Applied Biosystems brand. This system is for use with Applied Biosystems RapidLINK Software.
Features of the Applied Biosystems RapidHIT ID System include:
• Sample to answer in as little as 90 minutes
• 1-minute hands-on time with integrated ACE and RapidIntel PLUS sample cartridges
• Consumables tracking through radio frequency identification (RFID)
• Intuitive touchscreen interface
• Facial recognition and barcode camera
• Fingerprint reader
• Up to twelve months of shelf life for both sample and primary cartridges
• FBI NDIS-approved rapid DNA system and booking station device*
* NDIS-approved for use with RapidHIT ID ACE GlobalFiler Express Sample Cartridge in accredited forensic DNA laboratories
Specifications
Specifications
RapidHIT™
1 system
Room Temperature